Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144912.3 | NP_659161.2 | 5, 6 | 498 | ||
GenBank mRNA | AK136504.1 | - | 5, 6 | 534 | ||
AY297460.1 | - | 5, 6 | 498 | |||
BC021784.1 | - | 5, 6 | 498 | 4162418 |