Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rimbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081388.1 | NP_001074857.1 | 19 | 3551 | ||
GenBank mRNA | AK038814.1 | - | 18 | 3019 | ||
AK044685.1 | - | 18 | 3651 | |||
AK122245.2 | - | 10 | 2364 | |||
AK147417.1 | - | 15 | 2957 | |||
BC151100.1 | - | 15 | 3142 | 9088013 | ||
BC151101.1 | - | 15 | 3142 | 9088014 |