Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172725.2 | NP_766313.2 | 6 | 801 | ||
GenBank mRNA | AK040924.1 | - | 4 | 353 | ||
AK146431.1 | - | 6 | 679 | |||
AK152402.1 | - | 6 | 679 | |||
AK153787.1 | - | 6 | 674 | |||
AK181051.1 | - | 1 | 17 | |||
AK218835.1 | - | 3 | 147 | |||
BC050232.1 | - | 4 | 398 | |||
BC057548.1 | - | 6 | 656 | |||
BC082786.1 | - | 6 | 774 | |||
BC099419.1 | - | 6 | 733 | |||
BC144755.1 | - | 4 | 332 |