Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177682.3 | NP_808350.1 | 12 | 1120 | ||
GenBank mRNA | AK090008.1 | - | 12 | 1134 | ||
AK146704.1 | - | 12 | 1155 | |||
AK150793.1 | - | 12 | 1157 | |||
AK152328.1 | - | 12 | 1156 | |||
AK152576.1 | - | 12 | 1156 | |||
AK153160.1 | - | 12 | 1157 | |||
BC048077.1 | - | 12 | 1120 | 6308253 |