Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178058.1 | NP_001171529.1 | 8 | 1111 | ||
NM_153162.3 | NP_694802.2 | 8 | 1111 | |||
GenBank mRNA | AF349659.1 | - | 8 | 1020 | ||
AK012699.1 | - | 8 | 1104 | |||
AK041046.1 | - | 8 | 1114 | |||
AK080362.1 | - | 8 | 1102 | |||
AK135794.1 | - | 8 | 1058 | |||
AK205270.1 | - | 1 | 59 | |||
AK208532.1 | - | 3 | 314 | |||
BC076605.1 | - | 8 | 1069 | 30628949 |