Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172730.2 | NP_766318.2 | 8 | 833 | ||
GenBank mRNA | AK039818.1 | - | 8 | 812 | ||
AK046253.1 | - | 4 | 2679 | |||
AK081505.1 | - | 7 | 3478 | |||
AK156259.1 | - | 8 | 816 | |||
BC016607.1 | - | 6 | 601 | |||
BC051147.1 | - | 8 | 827 | 6595828 |