Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172731.2 | NP_766319.2 | 17 | 3835 | ||
GenBank mRNA | AK049341.1 | - | 4 | 509 | ||
AK086031.1 | - | 3 | 345 | |||
AK220153.1 | - | 18 | 4309 | |||
BC042732.1 | - | 13 | 1252 | |||
BC060664.1 | - | 17 | 3298 | |||
BC145502.1 | - | 18 | 3652 | 9054065 | ||
BC150786.1 | - | 17 | 3784 | 9087697 |