Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145148.2 | NP_660130.2 | 22 | 2626 | ||
GenBank mRNA | AB042027.1 | - | 22 | 2549 | ||
AF327856.1 | - | 21 | 2249 | |||
AF327857.1 | - | 20 | 2036 | |||
AK031491.1 | - | 11 | 1812 | |||
AK129262.1 | - | 21 | 2101 | |||
AK132280.1 | - | 22 | 2490 | |||
BC031369.1 | - | 6 | 740 | |||
BC039783.1 | - | 1 | 56 | |||
BC052390.1 | - | 22 | 2495 | 5718942 | ||
BC058262.1 | - | 11 | 1449 | 6487449 |