Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145148.2 | NP_660130.2 | 17 | 1883 | ||
GenBank mRNA | AB042027.1 | - | 17 | 1806 | ||
AF327856.1 | - | 16 | 1506 | |||
AF327857.1 | - | 15 | 1293 | |||
AK031491.1 | - | 6 | 1069 | |||
AK129262.1 | - | 16 | 1358 | |||
AK132280.1 | - | 17 | 1747 | |||
BC052390.1 | - | 17 | 1752 | 5718942 | ||
BC058262.1 | - | 6 | 706 | 6487449 |