Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172733.1 | NP_766321.1 | 1, 2 | 61 | ||
GenBank mRNA | AK012558.1 | - | 1, 2 | 97 | ||
AK030961.1 | - | 1, 2 | 93 | |||
AK034347.1 | - | 63 | ||||
AK075963.1 | - | 1, 2 | 86 | |||
AK083298.1 | - | 1, 2 | 61 | |||
AK145820.1 | - | 1, 2 | 97 | |||
AK150273.1 | - | 1, 2 | 87 | |||
AK150712.1 | - | 1, 2 | 57 | |||
AK152462.1 | - | 1, 2 | 65 | |||
AK168204.1 | - | 1, 2 | 97 | |||
AK169418.1 | - | 1, 2 | 81 | |||
BC016218.1 | - | 1, 2 | 67 | 4484163 |