Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175632.3 | NP_783440.1 | 1 | 105 | ||
GenBank mRNA | AF391161.1 | - | 1 | 81 |