Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001447.3 | NP_001001447.1 | 3 | 809 | ||
NM_001290436.1 | NP_001277365.1 | 3 | 846 | |||
NM_001290438.1 | NP_001277367.1 | 3 | 743 | |||
NM_001290440.1 | NP_001277369.1 | 2 | 258 | |||
NM_001290441.1 | NP_001277370.1 | 2 | 324 | |||
NM_001290442.1 | NP_001277371.1 | 2 | 361 | |||
GenBank mRNA | AK052559.1 | - | 3 | 804 | ||
AK078591.1 | - | 3 | 713 | |||
AK085207.1 | - | 3 | 804 | |||
AK163694.1 | - | 2 | 335 | |||
BB849089.1 | - | 2 | 259 | |||
BC026676.1 | - | 2 | 290 | 5053169 | ||
BY288068.1 | - | 2 | 314 |