Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004762.3 | NP_001004762.1 | 6, 7 | 789 | ||
NM_001168504.1 | NP_001161976.1 | 6, 7 | 805 | |||
GenBank mRNA | AK136098.1 | - | 6, 7 | 790 | ||
AK136143.1 | - | 6, 7 | 792 | |||
AK145339.1 | - | 6, 7 | 790 | |||
AK162196.1 | - | 6, 7 | 792 | |||
AY694793.1 | - | 6, 7 | 776 | |||
BC054740.1 | - | 4, 5 | 502 | |||
BC117808.1 | - | 6, 7 | 805 | 40092567 | ||
BC117809.1 | - | 6, 7 | 805 | 40092569 | ||
BC129974.1 | - | 6, 7 | 805 | 40092571 | ||
BC138548.1 | - | 6, 7 | 765 | 8861568 | ||
BC145767.1 | - | 6, 7 | 765 | 40131070 |