Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199149.3 | NP_954600.1 | 7 | 1193 | ||
GenBank mRNA | AK050946.1 | - | 7 | 1194 | ||
AK160935.1 | - | 7 | 1194 | |||
BC070402.1 | - | 7 | 1041 | 5696223 |