Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198027.2 | NP_932144.2 | 6 | 505 | ||
GenBank mRNA | AK082058.1 | - | 5 | 673 | ||
AK209174.1 | - | 2 | 153 | |||
BC029805.1 | - | 6 | 457 | 4952089 | ||
BC080310.1 | - | 4 | 628 |