Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146189.3 | NP_666301.2 | 16, 17 | 1818 | ||
GenBank mRNA | AK029282.1 | - | 16, 17 | 1804 | ||
AK132563.1 | - | 16, 17 | 1808 | |||
AK161146.1 | - | 16, 17 | 1806 | |||
BC029762.1 | - | 16, 17 | 1788 | 3983351 | ||
BC038869.1 | - | 1, 2 | 81 | |||
BC039184.1 | - | 16, 17 | 1783 | 3980667 |