Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146191.3 | NP_666303.3 | 31 | 5069 | ||
GenBank mRNA | AK040153.1 | - | 11 | 1751 | ||
AK076432.1 | - | 4 | 396 | |||
AK081849.1 | - | 1 | 62 | |||
AK129441.1 | - | 28 | 4482 | |||
AK147469.1 | - | 31 | 5055 | |||
AK154395.1 | - | 5 | 588 |