Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178070.4 | NP_835171.2 | 8, 9 | 937 | ||
GenBank mRNA | AK032076.1 | - | 8, 9 | 939 | ||
AK134927.1 | - | 8, 9 | 860 | |||
BC011059.1 | - | 8, 9 | 929 | |||
BC034170.1 | - | 8, 9 | 649 | 4971695 |