Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177695.4 | NP_808363.3 | 23 | 2873 | ||
GenBank mRNA | AY263157.1 | - | 23 | 2874 |