Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201352.2 | NP_958740.2 | 6 | 1175 | ||
GenBank mRNA | AK141189.1 | - | 6 | 1208 | ||
AK154771.1 | - | 6 | 1312 | |||
BC024955.1 | - | 5 | 461 | 4953174 | ||
BC026428.1 | - | 2 | 108 | 4224574 | ||
BC082585.1 | - | 6 | 1175 | 30608279 |