Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173739.3 | NP_776100.2 | 6 | 1437 | ||
GenBank mRNA | AB175685.1 | - | 6 | 1007 | ||
BC024988.1 | - | 6 | 1437 | 5044626 |