Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278943.1 | NP_001265872.1 | 14, 15 | 2201 | ||
NM_001278944.1 | NP_001265873.1 | 17, 18 | 2670 | |||
GenBank mRNA | AY243472.1 | - | 17, 18 | 2669 |