Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnrc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144925.3 | NP_659174.3 | 13, 14 | 4378 | ||
GenBank mRNA | AK044679.1 | - | 1, 2 | 762 | ||
AK138426.1 | - | 3, 4 | 428 | |||
AK147327.1 | - | 13, 14 | 4083 | |||
BC005741.1 | - | 2, 3 | 213 |