Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183020.1 | NP_898841.1 | 15, 16 | 2140 | ||
GenBank mRNA | AK155062.1 | - | 15, 16 | 2187 | ||
AK158626.1 | - | 15, 16 | 1827 | |||
AK168745.1 | - | 15, 16 | 1857 | |||
BC034083.1 | - | 9, 10 | 1219 | |||
BC043451.1 | - | 10, 11 | 1298 | |||
BC054483.1 | - | 15, 16 | 2140 | 6391591 |