Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sez6l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252566.1 | NP_001239495.1 | 16 | 3151 | ||
NM_001252567.1 | NP_001239496.1 | 17 | 3010 | |||
NM_144926.5 | NP_659175.1 | 17 | 3190 | |||
GenBank mRNA | AB206789.1 | - | 17 | 2791 | ||
AK034088.1 | - | 17 | 3162 | |||
AK165738.1 | - | 16 | 3123 | |||
BC011475.1 | - | 17 | 2996 | 4191879 | ||
BC079573.1 | - | 17 | 2579 | 5686330 | ||
BC096615.1 | - | 16 | 3063 | 6467632 |