Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172282.2 | NP_758486.1 | 7 | 1577 | ||
GenBank mRNA | AK033499.1 | - | 7 | 1338 | ||
AK046065.1 | - | 7 | 1367 | |||
AK049974.1 | - | 7 | 1336 | |||
AK088866.1 | - | 7 | 1361 | |||
AK148252.1 | - | 7 | 1380 | |||
BC039955.1 | - | 7 | 1323 | 4236297 |