Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpte | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199257.2 | NP_954866.2 | 14 | 1047 | ||
GenBank mRNA | AJ311311.1 | - | 14 | 1050 | ||
AJ311312.1 | - | 14 | 1135 | |||
AJ311313.1 | - | 14 | 1050 | |||
BC107329.2 | - | 14 | 842 | 40054415 |