Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpte | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199257.2 | NP_954866.2 | 14 | 1074 | ||
GenBank mRNA | AJ311311.1 | - | 14 | 1077 | ||
AJ311312.1 | - | 14 | 1162 | |||
AJ311313.1 | - | 14 | 1077 | |||
BC107329.2 | - | 14 | 869 | 40054415 |