Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145594.2 | NP_663569.2 | 4 | 420 | ||
GenBank mRNA | AF478470.1 | - | 4 | 430 | ||
AK083397.1 | - | 3 | 961 | |||
BC021946.1 | - | 4 | 388 | 5098805 | ||
BC029734.1 | - | 4 | 397 | 4158843 |