Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252623.1 | NP_001239552.1 | 8 | 2055 | ||
NM_172753.5 | NP_766341.4 | 7 | 1751 | |||
GenBank mRNA | AK028688.1 | - | 7 | 1674 | ||
AK033522.1 | - | 5 | 1405 | |||
AK080687.1 | - | 7 | 1720 | |||
AK133567.1 | - | 5 | 1404 | |||
AK144089.1 | - | 4 | 1312 | |||
BC026599.1 | - | 2 | 171 | |||
BC057630.1 | - | 9 | 2023 | |||
BC089162.1 | - | 7 | 1748 | 30470205 |