Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Colgalt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146211.3 | NP_666323.2 | 9 | 1402 | ||
GenBank mRNA | AK029433.1 | - | 9 | 1256 | ||
AK035788.1 | - | 9 | 1280 | |||
AK036165.1 | - | 9 | 1405 | |||
AK076103.1 | - | 9 | 1258 | |||
AK154775.1 | - | 9 | 1256 | |||
BC018287.1 | - | 9 | 1167 | |||
BC027801.1 | - | 2 | 289 | |||
BC032165.1 | - | 9 | 1434 | 5353334 | ||
BC034208.1 | - | 6 | 720 | |||
BC056951.1 | - | 9 | 1293 | 5695490 |