Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172757.3 | NP_766345.3 | 14 | 1949 | ||
GenBank mRNA | AK047782.1 | - | 14 | 1980 | ||
AK052635.1 | - | 14 | 1976 | |||
AK134510.1 | - | 14 | 1954 | |||
AK136424.1 | - | 14 | 1959 | |||
AK140522.1 | - | 14 | 1967 | |||
AK163533.1 | - | 1 | 25 | |||
AK163715.1 | - | 14 | 2712 | |||
AK165541.1 | - | 14 | 1843 | |||
BC030373.1 | - | 10 | 1294 | |||
BC048074.1 | - | 3 | 667 | |||
BC065060.1 | - | 14 | 1949 | 6402217 |