Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D230025D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145604.2 | NP_663579.2 | 9 | 1002 | ||
GenBank mRNA | AK051965.1 | - | 8 | 890 | ||
AK156120.1 | - | 9 | 1004 | |||
AK166937.1 | - | 8 | 2189 | |||
AK219810.1 | - | 1 | 49 | |||
BC006874.1 | - | 9 | 885 | 2646515 | ||
BC057919.1 | - | 9 | 991 | 5134302 | ||
DV649781.1 | - | 3 | 255 |