Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177449.3 | NP_803232.2 | 1, 2 | 48 | ||
GenBank mRNA | AK041606.1 | - | 1, 2 | 33 | ||
AK042435.1 | - | 1, 2 | 33 | |||
AK148120.1 | - | 1, 2 | 50 | |||
AK170400.1 | - | 1, 2 | 48 | |||
BC034290.1 | - | 1, 2 | 27 | 4038084 | ||
CB236245.1 | - | 1, 2 | 51 |