Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301100.1 | NP_001288029.1 | 26, 27 | 3845 | ||
NM_181594.3 | NP_853625.1 | 26, 27 | 3797 | |||
GenBank mRNA | AK146527.1 | - | 26, 27 | 3811 | ||
AK214755.1 | - | 1, 2 | 69 | |||
BC022641.1 | - | 5, 6 | 802 | |||
BC038076.1 | - | 5, 6 | 579 | |||
BC053081.1 | - | 26, 27 | 3727 | 5704584 |