Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146215.4 | NP_666327.2 | 2 | 1074 | ||
GenBank mRNA | AK032855.1 | - | 2 | 1050 | ||
AK050315.1 | - | 2 | 1046 | |||
AK158368.1 | - | 2 | 1056 | |||
BC025546.1 | - | 2 | 1056 | 5320939 |