Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vac14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146216.2 | NP_666328.2 | 13 | 1645 | ||
GenBank mRNA | AK031952.1 | - | 13 | 1680 | ||
AK043319.1 | - | 13 | 1677 | |||
AK046826.1 | - | 13 | 1617 | |||
AK054468.1 | - | 4 | 472 | |||
AK084520.1 | - | 13 | 1696 | |||
AK171577.1 | - | 13 | 1685 | |||
AK171883.1 | - | 13 | 1673 | |||
AK177604.1 | - | 2 | 82 | |||
AK183924.1 | - | 4 | 305 | |||
AK187577.1 | - | 2 | 82 | |||
AK191163.1 | - | 2 | 82 | |||
AK216581.1 | - | 2 | 83 | |||
BC032215.1 | - | 6 | 558 | |||
BC052199.1 | - | 13 | 1645 | 6512678 |