Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vac14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146216.2 | NP_666328.2 | 15 | 2058 | ||
GenBank mRNA | AK031952.1 | - | 15 | 2093 | ||
AK043319.1 | - | 15 | 2086 | |||
AK046826.1 | - | 15 | 2030 | |||
AK054468.1 | - | 6 | 885 | |||
AK084520.1 | - | 15 | 2109 | |||
AK140191.1 | - | 14 | 1928 | |||
AK171577.1 | - | 15 | 2098 | |||
AK171883.1 | - | 15 | 2086 | |||
AK179840.1 | - | 2 | 197 | |||
AK184343.1 | - | 2 | 197 | |||
AK184835.1 | - | 2 | 197 | |||
AK186210.1 | - | 2 | 198 | |||
AK220041.1 | - | 2 | 197 | |||
BC032215.1 | - | 8 | 971 | |||
BC052199.1 | - | 15 | 2058 | 6512678 |