Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172283.3 | NP_758487.2 | 7 | 642 | ||
GenBank mRNA | AJ297482.4 | - | 7 | 613 | ||
AJ534942.1 | - | 7 | 613 | |||
AK161763.1 | - | 7 | 643 | |||
AY223865.1 | - | 7 | 613 | |||
BC037698.1 | - | 6 | 591 | 4190449 |