Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172283.3 | NP_758487.2 | 21 | 2833 | ||
GenBank mRNA | AJ297482.4 | - | 21 | 2804 | ||
AJ534942.1 | - | 21 | 2804 | |||
AK049153.1 | - | 10 | 1662 | |||
AK131165.1 | - | 10 | 4183 | |||
AK161763.1 | - | 21 | 2937 | |||
AK164340.1 | - | 21 | 2900 | |||
AY223865.1 | - | 21 | 2804 | |||
BC037698.1 | - | 20 | 2782 | 4190449 |