Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190786.1 | NP_001177715.1 | 5 | 394 | ||
NM_001190800.1 | NP_001177729.1 | 5 | 437 | |||
NM_172284.5 | NP_758488.1 | 5 | 437 | |||
GenBank mRNA | AK013362.1 | - | 5 | 432 | ||
AK033235.1 | - | 5 | 395 | |||
AK147499.1 | - | 5 | 432 | |||
BC025594.1 | - | 5 | 397 | 5342859 |