Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181816.2 | NP_861537.2 | 8 | 1063 | ||
GenBank mRNA | AK030153.1 | - | 8 | 1045 | ||
AK077147.1 | - | 3 | 441 | |||
BC096547.1 | - | 7 | 956 | 30360184 | ||
KC211186.1 | - | 7 | 907 |