Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205367.1 | NP_001192296.1 | 12 | 1381 | ||
NM_009859.4 | NP_033989.2 | 13 | 1384 | |||
GenBank mRNA | AK040766.1 | - | 1 | 913 | ||
AK088997.1 | - | 13 | 1171 | |||
AK144127.1 | - | 13 | 1173 | |||
AK220447.1 | - | 11 | 3427 | |||
BC058587.1 | - | 12 | 1185 | 6529169 |