Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 11 | 1934 | ||
GenBank mRNA | AK028768.1 | - | 11 | 1569 | ||
AK030208.1 | - | 11 | 1566 | |||
AK032376.1 | - | 11 | 1000 | |||
AK033867.1 | - | 11 | 1430 | |||
AK051527.1 | - | 11 | 1935 | |||
AK078706.1 | - | 6 | 468 | |||
AK136229.1 | - | 6 | 459 | |||
BC023073.1 | - | 3 | 212 | |||
BC049909.1 | - | 11 | 1075 | |||
BC064100.1 | - | 1 | 50 | |||
BC094247.1 | - | 10 | 1419 | 5717984 |