Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gldn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177350.5 | NP_796324.1 | 8 | 1001 | ||
GenBank mRNA | AF548022.1 | - | 8 | 657 | ||
AK031523.1 | - | 8 | 1002 | |||
BC125025.1 | - | 8 | 981 | 40058551 | ||
BC125026.1 | - | 8 | 981 | 40058552 |