Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lctl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145835.2 | NP_665834.1 | 13 | 1826 | ||
GenBank mRNA | AF309072.1 | - | 13 | 1827 | ||
AK053515.1 | - | 12 | 1661 | |||
BC030631.1 | - | 9 | 1151 | 1195880 |