Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177352.4 | NP_796326.1 | 8, 9 | 786 | ||
GenBank mRNA | AK049275.1 | - | 8, 9 | 787 | ||
AK076293.1 | - | 8, 9 | 815 | |||
AK160477.1 | - | 8, 9 | 841 | |||
AK162119.1 | - | 8, 9 | 787 | |||
AK163031.1 | - | 8, 9 | 782 | |||
AK167217.1 | - | 8, 9 | 806 | |||
BC116302.1 | - | 8, 9 | 783 | 40095102 | ||
BC116303.1 | - | 7, 8 | 689 | 40095106 |