Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Topbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176979.5 | NP_795953.2 | 13 | 2134 | ||
GenBank mRNA | AK129104.1 | - | 13 | 2246 | ||
BC007170.1 | - | 8 | 1427 | |||
BC049797.1 | - | 13 | 2134 | 6510112 | ||
BC062111.1 | - | 2 | 198 |