Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145619.2 | NP_663594.2 | 8 | 1221 | ||
GenBank mRNA | AF368233.1 | - | 8 | 1221 | ||
AK145259.1 | - | 8 | 1255 | |||
AK147868.1 | - | 8 | 1520 | |||
AK165070.1 | - | 6 | 1470 | |||
AK170541.1 | - | 8 | 1165 |