Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camkv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145621.2 | NP_663596.1 | 5 | 550 | ||
GenBank mRNA | AK085143.1 | - | 556 | |||
AK147323.1 | - | 5 | 538 | |||
BC017634.1 | - | 5 | 550 | 4527857 | ||
BC022169.1 | - | 4 | 759 | |||
BC086762.1 | - | 4 | 748 | |||
BC111033.1 | - | 5 | 545 | 5368225 |